Fmoc-N-Me-Ile-OH 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR52601-25G
APOSOR52601-25GEA 75.6 GBP
APOSOR52601-25G
Fmoc-N-Me-Ile-OH 97%
Fmoc-N-Me-Ile-OH
Formula: C₂₂H₂₅NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00153389
CAS Number: 138775-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR