3,5-Dihydroxytoluene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR5254-100G OR5254-25G OR5254-500G
APOSOR5254-100GEA 135 GBP
APOSOR5254-100G APOSOR5254-25G APOSOR5254-500G
3,5-Dihydroxytoluene
3,5-Dihydroxytoluene
Formula: C₇H₈O₂
Boiling Pt: 287…290 °C
Melting Pt: 106…112 °C
Density: 1.29 g/cm³ (25 °C)
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00002291
CAS Number: 504-15-4
EINECS: 207-984-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR