Cyclohexanemethylamine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (Cyclohexylmethyl)amine, (Aminomethyl)cyclohexane, Cyclohexylmethanamine

OR5167-25G OR5167-100G
APOSOR5167-25GEA 137 GBP
APOSOR5167-25G APOSOR5167-100G
Cyclohexanemethylamine 95%
Cyclohexanemethylamine
Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 159…163 °C
Melting Pt: –8 °C
Density: 0.871 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00001520
CAS Number: 3218-02-8
EINECS: 221-741-8
UN: 2734
ADR: 8+3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR