7-Bromo-2-iodo-1-(phenylsulphonyl)-1H-indole 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR51177-1G OR51177-250MG
APOSOR51177-1GEA 473 GBP
APOSOR51177-1G APOSOR51177-250MG
7-Bromo-2-iodo-1-(phenylsulphonyl)-1H-indole 95%
7-Bromo-2-iodo-1-(phenylsulphonyl)-1H-indole
Formula: C₁₄H₉BrINO₂S
Storage Temperature: Ambient
CAS Number: 2514953-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR