6-Bromo-7-chloroimidazo[1,2-a]pyridine ≥97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR510043-250MG
APOSOR510043-250MGEA 64.1 GBP
APOSOR510043-250MG
6-Bromo-7-chloroimidazo[1,2-a]pyridine ≥97%
6-Bromo-7-chloroimidazo[1,2-a]pyridine
Formula: C₇H₄BrClN₂
MW: 231.477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405308
CAS Number: 1303890-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR