5-Bromo-2-chloro-1H-pyrrolo[2,3-b]pyridine ≥97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR510042-250MG
APOSOR510042-250MGEA 153 GBP
APOSOR510042-250MG
5-Bromo-2-chloro-1H-pyrrolo[2,3-b]pyridine ≥97%
5-Bromo-2-chloro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₄BrClN₂
MW: 231.477 g/mol
Storage Temperature: Refrigerator
CAS Number: 1612287-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR