2-Chloro-3-methoxybenzonitrile ≥98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR510010-1G
APOSOR510010-1GEA 87.3 GBP
APOSOR510010-1G
2-Chloro-3-methoxybenzonitrile ≥98%
2-Chloro-3-methoxybenzonitrile
Formula: C₈H₆ClNO
Storage Temperature: Refrigerator
CAS Number: 853331-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR