6-Chloronicotinic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 6-Chloropyridine-3-carboxylic acid, 6-Chloro-3-pyridinecarboxylic acid

OR5069-100G OR5069-25G OR5069-500G
APOSOR5069-100GEA 31.5 GBP
APOSOR5069-100G APOSOR5069-25G APOSOR5069-500G
6-Chloronicotinic acid 98%
6-Chloronicotinic acid
Formula: C₆H₄ClNO₂
Melting Pt: 190…192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006241
CAS Number: 5326-23-8
EINECS: 226-201-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR