(1-Ethyl-3-methyl-1H-pyrazol-4-yl)methylamine 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480852-1G
APOSOR480852-1GEA 277 GBP
APOSOR480852-1G
(1-Ethyl-3-methyl-1H-pyrazol-4-yl)methylamine 95%
(1-Ethyl-3-methyl-1H-pyrazol-4-yl)methylamine
Formula: C₇H₁₃N₃
Storage Temperature: Ambient
MDL Number: MFCD02090911
CAS Number: 848436-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR