Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480832-1G OR480832-5G
APOSOR480832-1GEA 75.6 GBP
APOSOR480832-1G APOSOR480832-5G
Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate 95%
Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate
Formula: C₇H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09759206
CAS Number: 3697-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR