5-Chloro-1H-indole-3-carboxylic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480765-5G
APOSOR480765-5GEA 101 GBP
APOSOR480765-5G
5-Chloro-1H-indole-3-carboxylic acid 95%
5-Chloro-1H-indole-3-carboxylic acid
Formula: C₉H₆ClNO₂
MW: 195.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03410308
CAS Number: 10406-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR