(2R,4S)-4-Hydroxypiperidine-2-carboxylic acid 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480267-1G OR480267-250MG OR480267-500MG
APOSOR480267-1GEA 625 GBP
APOSOR480267-1G APOSOR480267-250MG APOSOR480267-500MG
(2R,4S)-4-Hydroxypiperidine-2-carboxylic acid 95%
(2R,4S)-4-Hydroxypiperidine-2-carboxylic acid
Formula: C₆H₁₁NO₃·ClH
MW: 181.617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110198
CAS Number: 175671-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR