Ethyl 5-amino-1-(2-methoxyphenyl)pyrazole-3-carboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480050-1G
APOSOR480050-1GEA 577 GBP
APOSOR480050-1G
Ethyl 5-amino-1-(2-methoxyphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(2-methoxyphenyl)pyrazole-3-carboxylate
Formula: C₁₃H₁₅N₃O₃
MW: 261.277 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07388421
CAS Number: 1427012-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR