(R)-1-Cbz-3-(Hydroxymethyl)pyrrolidine 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (R)-1-Cbz-3-(hydroxymethyl)pyrrolidine

OR471682-1G
APOSOR471682-1GEA 64.1 GBP
APOSOR471682-1G
(R)-1-Cbz-3-(Hydroxymethyl)pyrrolidine 97%
(R)-1-Cbz-3-(Hydroxymethyl)pyrrolidine
Formula: C₁₃H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD06410562
CAS Number: 192214-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR