3-(Cbz-aminomethyl)phenol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471537-25G OR471537-5G
APOSOR471537-25GEA 729 GBP
APOSOR471537-25G APOSOR471537-5G
3-(Cbz-aminomethyl)phenol
3-(Cbz-aminomethyl)phenol
Formula: C₁₅H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD14585006
CAS Number: 75383-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR