(E)-4-(Dimethylamino)-N-[4-(hydroxymethyl)phenyl]-2-butenamide hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471514-250MG
APOSOR471514-250MGEA 516 GBP
APOSOR471514-250MG
(E)-4-(Dimethylamino)-N-[4-(hydroxymethyl)phenyl]-2-butenamide hydrochloride
(E)-4-(Dimethylamino)-N-[4-(hydroxymethyl)phenyl]-2-butenamide hydrochloride
Formula: C₁₃H₁₈N₂O₂·ClH
MW: 270.755 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29049240
CAS Number: 2006281-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR