1-[2-(1-Piperidyl)phenyl]ethanol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471439-1G OR471439-5G
APOSOR471439-1GEA 435 GBP
APOSOR471439-1G APOSOR471439-5G
1-[2-(1-Piperidyl)phenyl]ethanol
1-[2-(1-Piperidyl)phenyl]ethanol
Formula: C₁₃H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD12187296
CAS Number: 78648-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR