3-(Aminomethyl)-1-tosylpyrrole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471260-1G
APOSOR471260-1GEA 1110 GBP
APOSOR471260-1G
3-(Aminomethyl)-1-tosylpyrrole
3-(Aminomethyl)-1-tosylpyrrole
Formula: C₁₂H₁₄N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD28398151
CAS Number: 872405-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR