2,4,6-Triiodophenol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR4709-25G OR4709-5G OR4709-100G
APOSOR4709-25GEA 89.8 GBP
APOSOR4709-25G APOSOR4709-5G APOSOR4709-100G
2,4,6-Triiodophenol
2,4,6-Triiodophenol
Formula: C₆H₃I₃O
Storage Temperature: Ambient
MDL Number: MFCD00002179
CAS Number: 609-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR