2-Bromo-5-iodothiophene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR470871-5G
APOSOR470871-5GEA 516 GBP
APOSOR470871-5G
2-Bromo-5-iodothiophene
2-Bromo-5-iodothiophene
Formula: C₄H₂BrIS
MW: 288.932 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08695058
CAS Number: 29504-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR