4-(3-Piperidyl)-1-butanol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR470564-5G
APOSOR470564-5GEA 662 GBP
APOSOR470564-5G
4-(3-Piperidyl)-1-butanol
4-(3-Piperidyl)-1-butanol
Formula: C₉H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD23381096
CAS Number: 1021910-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR