(R)-(-)-1-Cbz-3-Pyrrolidinol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR470000-25G
APOSOR470000-25GEA 221 GBP
APOSOR470000-25G
(R)-(-)-1-Cbz-3-Pyrrolidinol
(R)-(-)-1-Cbz-3-Pyrrolidinol
Formula: C₁₂H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD07368258
CAS Number: 100858-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR