1-Bicyclo[4.2.0]octa-1,3,5-trien-7-ylmethanamine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR46644-1G
APOSOR46644-1GEA 326 GBP
APOSOR46644-1G
1-Bicyclo[4.2.0]octa-1,3,5-trien-7-ylmethanamine
1-Bicyclo[4.2.0]octa-1,3,5-trien-7-ylmethanamine
Formula: C₉H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD06213241
CAS Number: 1005-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR