Triethylene glycol 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol

OR46604-10L OR46604-2.5L OR46604-500ML
APOSOR46604-10LEA 262 GBP
APOSOR46604-10L APOSOR46604-2.5L APOSOR46604-500ML
Triethylene glycol 99%
Triethylene glycol
Formula: C₆H₁₄O₄
Boiling Pt: 284…288 °C
Melting Pt: –7 °C
Density: 1.125 g/cm³ (25 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002880
CAS Number: 112-27-6
EINECS: 203-953-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR