(S)-(+)-2-(Methoxymethyl)pyrrolidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (S)-2-(Methoxymethyl)pyrrolidine

OR4615-1G OR4615-250MG
APOSOR4615-1GEA 27.3 GBP
APOSOR4615-1G APOSOR4615-250MG
(S)-(+)-2-(Methoxymethyl)pyrrolidine
(S)-(+)-2-(Methoxymethyl)pyrrolidine
Formula: C₆H₁₃NO
Boiling Pt: 61…62 °C/40mmHg
Density: 0.932 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00010408
CAS Number: 63126-47-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR