Tridecanol (mixture of isomers)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR46094-10G OR46094-1G
APOSOR46094-10GEA 17.9 GBP
APOSOR46094-10G APOSOR46094-1G
Tridecanol (mixture of isomers)
Tridecanol (mixture of isomers)
Formula: C₁₃H₂₈O
Boiling Pt: 155…156 °C/15mmHg
Melting Pt: 29…34 °C
Density: 0.822 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01778419
CAS Number: 26248-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR