2-Tolualdehyde 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: o-Tolualdehyde, 2-Methylbenzaldehyde

OR46026-100G OR46026-1KG OR46026-500G OR46026-25G
APOSOR46026-100GEA 31.5 GBP
APOSOR46026-100G APOSOR46026-1KG APOSOR46026-500G APOSOR46026-25G
2-Tolualdehyde 98%
2-Tolualdehyde
Formula: C₈H₈O
Boiling Pt: 199…200 °C
Melting Pt: –35 °C
Density: 1.037 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00003338
CAS Number: 529-20-4
EINECS: 208-452-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR