(R)-(-)-3-Aminopiperidine dihydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (R)-(-)-3-Piperidinamine dihydrochloride, (R)-Piperidin-3-amine dihydrochloride

OR4587-1G
APOSOR4587-1GEA 12.7 GBP
APOSOR4587-1G
(R)-(-)-3-Aminopiperidine dihydrochloride
(R)-(-)-3-Aminopiperidine dihydrochloride

Chiral intermediate for pharmaceuticals and agrochemicals.

Formula: C₅H₁₄Cl₂N₂
MW: 173.08 g/mol
Melting Pt: 190…195 °C
Storage Temperature: Ambient
MDL Number: MFCD06799458
CAS Number: 334618-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR