N-Benzylphenethylamine 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR451330-100G OR451330-250G OR451330-25G
APOSOR451330-100GEA 221 GBP
APOSOR451330-100G APOSOR451330-250G APOSOR451330-25G
N-Benzylphenethylamine 99%
N-Benzylphenethylamine
Formula: C₁₅H₁₇N
Boiling Pt: 162…165 °C
Density: 1.007 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009624
CAS Number: 3647-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR