Boc-2-Cyclopentylglycine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR451211-1G
APOSOR451211-1GEA 479 GBP
APOSOR451211-1G
Boc-2-Cyclopentylglycine
Boc-2-Cyclopentylglycine
Formula: C₁₂H₂₁NO₄
MW: 243.299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11707034
CAS Number: 35264-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR