(R)-3-Benzyloxypyrrolidine hydrochloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR451182-5G
APOSOR451182-5GEA 579 GBP
APOSOR451182-5G
(R)-3-Benzyloxypyrrolidine hydrochloride
(R)-3-Benzyloxypyrrolidine hydrochloride
Formula: C₁₁H₁₅NO·ClH
Storage Temperature: Ambient
MDL Number: MFCD09261313
CAS Number: 927819-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR