1,4-Dioxaspiro[4.5]decane-8-methanol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1,4-Dioxaspiro[4.5]decan-8-ylmethanol

OR451159-1G OR451159-250MG OR451159-5G OR451159-10G
APOSOR451159-1GEA 30.5 GBP
APOSOR451159-1G APOSOR451159-250MG APOSOR451159-5G APOSOR451159-10G
1,4-Dioxaspiro[4.5]decane-8-methanol
1,4-Dioxaspiro[4.5]decane-8-methanol
Formula: C₉H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD07779441
CAS Number: 17159-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR