(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

OR43631-1G OR43631-5G
APOSOR43631-1GEA 15.8 GBP
APOSOR43631-1G APOSOR43631-5G
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
Melting Pt: 239…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR