2-Chloro-3-nitro-5-thiophenesulphonyl chloride

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR4069-1G
APOSOR4069-1GEA 21 GBP
APOSOR4069-1G
2-Chloro-3-nitro-5-thiophenesulphonyl chloride
2-Chloro-3-nitro-5-thiophenesulphonyl chloride
Formula: C₄HCl₂NO₄S₂
Melting Pt: 51…54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067978
CAS Number: 58457-24-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR