Dimethylcyclopropane-1,1-dicarboxylate 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR40111-1G
APOSOR40111-1GEA 15.8 GBP
APOSOR40111-1G
Dimethylcyclopropane-1,1-dicarboxylate 99%
Dimethylcyclopropane-1,1-dicarboxylate
Formula: C₇H₁₀O₄
Boiling Pt: 196…198 °C
Density: 1.147 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00192079
CAS Number: 6914-71-2
EINECS: 414-240-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR