3-Chloro-4-methoxybenzyl bromide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4-(Bromomethyl)-2-chloro-1-methoxybenzene

OR400991-5G
APOSOR400991-5GEA 124 GBP
APOSOR400991-5G
3-Chloro-4-methoxybenzyl bromide
3-Chloro-4-methoxybenzyl bromide
Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD06858766
CAS Number: 320407-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR