2-Bromo-3-nitrobenzyl bromide

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

OR400520-1G OR400520-250MG OR400520-5G
APOSOR400520-1GEA 42 GBP
APOSOR400520-1G APOSOR400520-250MG APOSOR400520-5G
2-Bromo-3-nitrobenzyl bromide
2-Bromo-3-nitrobenzyl bromide
Formula: C₇H₅Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD18390263
CAS Number: 82617-49-0
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR