α,4-Dichlorotoluene 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 1-Chloro-4-(chloromethyl)benzene, 4-Chlorobenzyl chloride

OR3764-1KG OR3764-250G OR3764-5KG OR3764-25G OR3764-100G OR3764-400G
APOSOR3764-1KGEA 74.6 GBP
APOSOR3764-1KG APOSOR3764-250G APOSOR3764-5KG APOSOR3764-25G APOSOR3764-100G APOSOR3764-400G
α,4-Dichlorotoluene 99%
α,4-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 216…222 °C
Melting Pt: 26…30 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00000914
CAS Number: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR