2-Quinoxalinecarboxylic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR3738-10G OR3738-5G OR3738-25G
APOSOR3738-10GEA 29.4 GBP
APOSOR3738-10G APOSOR3738-5G APOSOR3738-25G
2-Quinoxalinecarboxylic acid
2-Quinoxalinecarboxylic acid
Formula: C₉H₆N₂O₂
Melting Pt: 208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012334
CAS Number: 879-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR