6-Bromoquinolin-4-ol

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 6-Bromo-4-quinolinol, 6-Bromo-4-hydroxyquinoline

OR3631-1G OR3631-5G
APOSOR3631-1GEA 0 GBP
APOSOR3631-1G APOSOR3631-5G
6-Bromoquinolin-4-ol
6-Bromoquinolin-4-ol
Formula: C₉H₆BrNO
Melting Pt: 281…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00272435
CAS Number: 145369-94-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR