N-Methylpentan-2-amine hydrochloride ≥98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR361721-500MG
APOSOR361721-500MGEA 247 GBP
APOSOR361721-500MG
N-Methylpentan-2-amine hydrochloride ≥98%
N-Methylpentan-2-amine hydrochloride
Formula: C₆H₁₅N·ClH
MW: 137.651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11099634
CAS Number: 130985-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR