4-(tert-Butoxycarbonyl-n-methylamino)-phenylboronic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (4-((tert-Butoxycarbonyl)(methyl)amino)phenyl)boronic acid

OR361605-1G OR361605-5G
APOSOR361605-1GEA 83.9 GBP
APOSOR361605-1G APOSOR361605-5G
4-(tert-Butoxycarbonyl-n-methylamino)-phenylboronic acid 98%
4-(tert-Butoxycarbonyl-n-methylamino)-phenylboronic acid
Formula: C₁₂H₁₈BNO₄
Storage Temperature: Ambient
MDL Number: MFCD03095105
CAS Number: 945756-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR