4-(2-Hydroxyethyl)phenylboronic acid 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (4-(2-Hydroxyethyl)phenyl)boronic acid

OR361251-250MG
APOSOR361251-250MGEA 31.5 GBP
APOSOR361251-250MG
4-(2-Hydroxyethyl)phenylboronic acid 97%
4-(2-Hydroxyethyl)phenylboronic acid
Formula: C₈H₁₁BO₃
MW: 165.982 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095135
CAS Number: 137756-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR