(3,4,5-Trimethoxyphenyl)acetonitrile

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 3,4,5-Trimethoxyphenylacetonitrile

OR350379-100G
APOSOR350379-100GEA 357 GBP
APOSOR350379-100G
(3,4,5-Trimethoxyphenyl)acetonitrile
(3,4,5-Trimethoxyphenyl)acetonitrile
Formula: C₁₁H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD00001912
CAS Number: 13338-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR