3-Nitro-o-xylene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR350286-500G OR350286-1KG OR350286-250G
APOSOR350286-500GEA 94.9 GBP
APOSOR350286-500G APOSOR350286-1KG APOSOR350286-250G
3-Nitro-o-xylene
3-Nitro-o-xylene
Formula: C₈H₉NO₂
Boiling Pt: 244…246 °C
Melting Pt: 13…15 °C
Density: 1.2129 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00007162
CAS Number: 83-41-0
UN: 1665
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR