Methyl (S)-(+)-2-pyrrolidone-5-carboxylate

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR350259-500G
APOSOR350259-500GEA 74.6 GBP
APOSOR350259-500G
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate
Formula: C₆H₉NO₃
Boiling Pt: 90 °C/ 0.1mmHg
Density: 1.226 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00080931
CAS Number: 4931-66-2
EINECS: 225-567-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR