5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Methyl 5-hydroxy-1-methyl-1H-pyrazole-3-carboxylate

OR346186-5G
APOSOR346186-5GEA 79.8 GBP
APOSOR346186-5G
5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester
5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester
Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01851055
CAS Number: 51985-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR