5-Bromo-2-chloromethyl-1H-benzoimidazole

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 5-Bromo-2-(chloromethyl)-1H-benzo[d]imidazole

OR346101-1G OR346101-5G OR346101-10G
APOSOR346101-1GEA 0 GBP
APOSOR346101-1G APOSOR346101-5G APOSOR346101-10G
5-Bromo-2-chloromethyl-1H-benzoimidazole
5-Bromo-2-chloromethyl-1H-benzoimidazole
Formula: C₈H₆BrClN₂
MW: 245.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00457491
CAS Number: 1740-88-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR