4'-(Methylthio)acetophenone

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR3455-100G OR3455-25G OR3455-5G
APOSOR3455-100GEA 63 GBP
APOSOR3455-100G APOSOR3455-25G APOSOR3455-5G
4'-(Methylthio)acetophenone
4'-(Methylthio)acetophenone
Formula: C₉H₁₀OS
Boiling Pt: 135 °C/1mmHg
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00039835
CAS Number: 1778-09-2
EINECS: 217-213-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR