(2-Ethyl-4-methoxy)phenylboronic acid

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

OR345427-250MG
APOSOR345427-250MGEA 294 GBP
APOSOR345427-250MG
(2-Ethyl-4-methoxy)phenylboronic acid
(2-Ethyl-4-methoxy)phenylboronic acid
Formula: C₉H₁₃BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD22414595
CAS Number: 852946-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR